Assay Details
| Gene Symbol: Zfp266 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082485.1 | NP_001075954.1 | 3 - 4 | 345 | 101 | |
| NM_001135019.1 | NP_001128491.1 | 3 - 4 | 334 | 101 | ||
| GenBank mRNA | AK019992.1 | - | 3 - 4 | 330 | 101 | |
| BC048701.1 | - | 3 - 4 | 285 | 101 | ||
| BC061209.1 | - | 3 - 4 | 331 | 101 | ||
| BC089552.1 | - | 3 - 4 | 344 | 101 | ||
| CJ053742.1 | - | 3 - 4 | 346 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC061209.1, NM_001082485.1, NM_001135019.1 transcripts.
Set Membership: |
|

Zfp266
Genomic Map