Assay Details
| Gene Symbol: Ttc12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172770.3 | NP_766358.1 | 6 - 7 | 536 | 67 | |
| GenBank mRNA | AK054350.1 | - | 6 - 7 | 537 | 67 | |
| AK076457.1 | - | 5 - 6 | 1027 | 67 | ||
| BC056616.1 | - | 5 - 6 | 464 | 4974294 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ttc12
Genomic Map