Assay Details
| Gene Symbol: Fank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025850.2 | NP_080126.1 | 2 - 3 | 257 | 65 | |
| GenBank mRNA | AK005694.1 | - | 2 - 3 | 267 | 65 | |
| AK017071.1 | - | 2 - 3 | 281 | 65 | ||
| AY819025.1 | - | 2 - 3 | 287 | 65 | ||
| BC046278.1 | - | 2 - 3 | 257 | 6310911 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fank1
Genomic Map