Assay Details
| Gene Symbol: Fank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025850.2 | NP_080126.1 | 3 - 4 | 383 | 91 | |
| GenBank mRNA | AK005694.1 | - | 3 - 4 | 393 | 91 | |
| AK017071.1 | - | 3 - 4 | 407 | 91 | ||
| AY819025.1 | - | 3 - 4 | 413 | 91 | ||
| BC046278.1 | - | 3 - 4 | 383 | 6310911 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fank1
Genomic Map