Assay Details
| Gene Symbol: Fank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025850.2 | NP_080126.1 | 7 - 8 | 771 | 72 | |
| GenBank mRNA | AK005694.1 | - | 7 - 8 | 781 | 72 | |
| AK017071.1 | - | 7 - 8 | 795 | 72 | ||
| AY819025.1 | - | 7 - 8 | 801 | 72 | ||
| BC046278.1 | - | 7 - 8 | 771 | 6310911 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fank1
Genomic Map