Assay Details
| Gene Symbol: Fank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025850.2 | NP_080126.1 | 9 - 10 | 997 | 126 | |
| GenBank mRNA | AY819025.1 | - | 9 - 10 | 1027 | 126 | |
| BC046278.1 | - | 9 - 10 | 997 | 6310911 | 126 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fank1
Genomic Map