Assay Details
| Gene Symbol: Stard3nl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK002760.1 | - | 6 - 7 | 734 | 94 | |
| AK152116.1 | - | 7 - 8 | 797 | 94 | ||
| AK152378.1 | - | 7 - 8 | 748 | 94 | ||
| BC003334.1 | - | 7 - 8 | 751 | 3256725 | 94 | |
| CO039188.1 | - | 2 - 3 | 94 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Stard3nl
Genomic Map