Assay Details
| Gene Symbol: Cdh18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081299.1 | NP_001074768.1 | 4 - 5 | 1011 | 106 | |
| GenBank mRNA | AK045672.1 | - | 4 - 5 | 1163 | 106 | |
| AK137430.1 | - | 4 - 5 | 1011 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdh18
Genomic Map