Assay Details
| Gene Symbol: Wnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023653.5 | NP_076142.3 | 3 - 4 | 738 | 80 | |
| GenBank mRNA | AK012093.1 | - | 3 - 4 | 654 | 80 | |
| AK045120.1 | - | 3 - 4 | 650 | 80 | ||
| AK167642.1 | - | 3 - 4 | 654 | 80 | ||
| BC026373.1 | - | 3 - 4 | 633 | 4162686 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wnt2
Genomic Map