Assay Details
| Gene Symbol: Ccdc132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167750.1 | NP_001161222.1 | 24 - 25 | 2439 | 61 | |
| NM_001167751.1 | NP_001161223.1 | 24 - 25 | 2439 | 61 | ||
| NM_024260.5 | NP_077222.4 | 24 - 25 | 2439 | 61 | ||
| GenBank mRNA | AK006602.1 | - | 6 - 7 | 665 | 61 | |
| AK042031.1 | - | 24 - 25 | 2419 | 61 | ||
| AK075898.1 | - | 24 - 25 | 2428 | 61 | ||
| AK129458.1 | - | 24 - 25 | 2389 | 61 | ||
| AK134533.1 | - | 8 - 9 | 4238 | 61 | ||
| AK217072.1 | - | 2 - 3 | 236 | 61 | ||
| AY254697.1 | - | 24 - 25 | 2432 | 61 | ||
| BC002304.1 | - | 9 - 10 | 984 | 3591600 | 61 | |
| BC036294.1 | - | 24 - 25 | 2379 | 3500316 | 61 | |
| BC043092.1 | - | 24 - 25 | 2396 | 6405356 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc132
Genomic Map