Assay Details
| Gene Symbol: Rps6ka3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148945.2 | NP_683747.1 | 6 - 7 | 825 | 130 | |
| GenBank mRNA | AK079102.1 | - | 918 | 130 | ||
| AK154778.1 | - | 6 - 7 | 738 | 130 | ||
| AY083469.1 | - | 6 - 7 | 487 | 130 | ||
| BC150156.1 | - | 6 - 7 | 490 | 130 | ||
| BC150478.1 | - | 6 - 7 | 490 | 130 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rps6ka3
Genomic Map