| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300009803.mm.1 |
NM_001251833.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009803.mm.1 |
NM_001251833.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009803.mm.1 |
NM_001251834.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009803.mm.1 |
NM_001251834.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009803.mm.1 |
NM_139141.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009803.mm.1 |
NM_139141.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009803.mm.1 |
TC1300001547.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300009809.mm.1 |
NM_001251833.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009809.mm.1 |
NM_001251833.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009809.mm.1 |
NM_001251834.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009809.mm.1 |
NM_001251834.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009809.mm.1 |
NM_139141.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009809.mm.1 |
NM_139141.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009809.mm.1 |
TC1300001547.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300009804.mm.1 |
NM_001251833.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009804.mm.1 |
NM_001251833.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009804.mm.1 |
NM_001251834.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009804.mm.1 |
NM_001251834.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009804.mm.1 |
NM_139141.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009804.mm.1 |
NM_139141.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009804.mm.1 |
TC1300001547.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300009808.mm.1 |
NM_001251833.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009808.mm.1 |
NM_001251833.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009808.mm.1 |
NM_001251834.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009808.mm.1 |
NM_001251834.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009808.mm.1 |
NM_139141.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009808.mm.1 |
NM_139141.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009808.mm.1 |
TC1300001547.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300009807.mm.1 |
NM_001251833.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009807.mm.1 |
NM_001251833.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009807.mm.1 |
NM_001251834.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009807.mm.1 |
NM_001251834.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009807.mm.1 |
NM_139141.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009807.mm.1 |
NM_139141.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009807.mm.1 |
TC1300001547.mm.1 |
MTA TC |