Assay Details
| Gene Symbol: Zkscan8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251833.1 | NP_001238762.1 | 2 - 3 | 670 | 65 | |
| NM_139141.4 | NP_631880.2 | 2 - 3 | 670 | 65 | ||
| GenBank mRNA | AK032718.1 | - | 2 - 3 | 626 | 65 | |
| AK134299.1 | - | 2 - 3 | 655 | 65 | ||
| BC050152.1 | - | 2 - 3 | 606 | 65 | ||
| BC145448.1 | - | 1 - 2 | 504 | 9054006 | 65 | |
| BC150775.1 | - | 1 - 2 | 504 | 9087686 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zkscan8
Genomic Map