Assay Details
| Gene Symbol: AW554918 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033532.3 | NP_001028704.2 | 3 - 4 | 247 | 69 | |
| GenBank mRNA | AK167257.1 | - | 3 - 4 | 248 | 69 | |
More Information
Set Membership: |
|

AW554918
Genomic Map