Assay Details
| Gene Symbol: Aoah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012054.4 | NP_036184.1 | 9 - 10 | 978 | 101 | |
| GenBank mRNA | AK084452.1 | - | 9 - 10 | 973 | 101 | |
| AK085343.1 | - | 9 - 10 | 950 | 101 | ||
| AK133301.1 | - | 9 - 10 | 919 | 101 | ||
| AK170293.1 | - | 8 - 9 | 514 | 101 | ||
| AK171572.1 | - | 9 - 10 | 797 | 101 | ||
| BC150767.1 | - | 9 - 10 | 772 | 9087678 | 101 | |
| CT010242.1 | - | 9 - 10 | 697 | 101 | ||
More Information
Set Membership: |
|

Aoah
Genomic Map