Assay Details
| Gene Symbol: Cdh10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009865.2 | NP_033995.1 | 2 - 3 | 715 | 134 | |
| GenBank mRNA | AF183946.1 | - | 2 - 3 | 484 | 134 | |
| AK043661.1 | - | 2 - 3 | 458 | 134 | ||
| AK138606.1 | - | 2 - 3 | 717 | 134 | ||
| BC052056.1 | - | 2 - 3 | 423 | 5696194 | 134 | |
| BC062962.1 | - | 2 - 3 | 423 | 5696194 | 134 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdh10
Genomic Map