| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600000749.mm.1 |
NM_175088.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000749.mm.1 |
NM_175088.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000749.mm.1 |
TC0600000115.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600000746.mm.1 |
NM_175088.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000746.mm.1 |
NM_175088.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000746.mm.1 |
TC0600000115.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600000752.mm.1 |
NM_175088.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000752.mm.1 |
NM_175088.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000752.mm.1 |
TC0600000115.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600000753.mm.1 |
NM_175088.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000753.mm.1 |
NM_175088.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000753.mm.1 |
TC0600000115.mm.1 |
MTA TC |