Assay Details
| Gene Symbol: Kif20a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166406.1 | NP_001159878.1 | 13 - 14 | 1933 | 80 | |
| NM_001166407.1 | NP_001159879.1 | 13 - 14 | 1931 | 80 | ||
| NM_009004.4 | NP_033030.1 | 13 - 14 | 1827 | 80 | ||
| GenBank mRNA | AK051201.1 | - | 13 - 14 | 1745 | 80 | |
| AK051995.1 | - | 13 - 14 | 1782 | 80 | ||
| AK053280.1 | - | 13 - 14 | 1746 | 80 | ||
| AK083412.1 | - | 13 - 14 | 1787 | 80 | ||
| AK083697.1 | - | 13 - 14 | 1748 | 80 | ||
| AK084732.1 | - | 13 - 14 | 1769 | 80 | ||
| AK142764.1 | - | 13 - 14 | 1775 | 80 | ||
| AK144844.1 | - | 13 - 14 | 1745 | 80 | ||
| BC060608.1 | - | 13 - 14 | 1933 | 5720408 | 80 | |
| Y09632.1 | - | 13 - 14 | 1982 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kif20a
Genomic Map