Assay Details
| Gene Symbol: Calu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_184053.3 | NP_908942.1 | 3 - 4 | 525 | 87 | |
| GenBank mRNA | AK147756.1 | - | 3 - 4 | 478 | 87 | |
| AK166705.1 | - | 3 - 4 | 478 | 87 | ||
| AY225334.1 | - | 3 - 4 | 451 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Calu
Genomic Map