Assay Details
| Gene Symbol: Stim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009287.4 | NP_033313.2 | 7 - 8 | 1560 | 82 | |
| GenBank mRNA | AK041944.1 | - | 7 - 8 | 1579 | 82 | |
| BC021644.1 | - | 7 - 8 | 1562 | 4191156 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stim1
Genomic Map