Assay Details
| Gene Symbol: Stim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009287.4 | NP_033313.2 | 8 - 9 | 1726 | 116 | |
| GenBank mRNA | AK041944.1 | - | 8 - 9 | 1745 | 116 | |
| BC021644.1 | - | 8 - 9 | 1728 | 4191156 | 116 | |
| U47323.1 | - | 8 - 9 | 1328 | 116 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stim1
Genomic Map