Assay Details
| Gene Symbol: Tmco6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028036.3 | NP_082312.3 | 10 - 11 | 1306 | 74 | |
| GenBank mRNA | AK010487.1 | - | 10 - 11 | 1231 | 74 | |
| BC005637.1 | - | 10 - 11 | 1308 | 3156734 | 74 | |
| BF468256.2 | - | 2 - 3 | 174 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmco6
Genomic Map