Assay Details
| Gene Symbol: F420014N23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045715.1 | 1 - 2 | 275 | 92 | ||
| GenBank mRNA | AK077682.1 | - | 1 - 2 | 276 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

F420014N23Rik
Genomic Map