Assay Details
| Gene Symbol: F420014N23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045715.1 | 2 - 3 | 377 | 79 | ||
| GenBank mRNA | AK077682.1 | - | 2 - 3 | 378 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

F420014N23Rik
Genomic Map