Assay Details
| Gene Symbol: Prss58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175020.3 | NP_778185.1 | 1 - 2 | 113 | 110 | |
| GenBank mRNA | AK075754.1 | - | 1 - 2 | 114 | 110 | |
| BC048599.1 | - | 1 - 2 | 107 | 6740675 | 110 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK075754.1, BC048599.1, NM_175020.3 transcripts.
Set Membership: |
|

Prss58
Genomic Map