Assay Details
| Gene Symbol: Prss58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175020.3 | NP_778185.1 | 4 - 5 | 603 | 59 | |
| GenBank mRNA | AK075754.1 | - | 4 - 5 | 604 | 59 | |
| BC048599.1 | - | 4 - 5 | 597 | 6740675 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prss58
Genomic Map