Assay Details
| Gene Symbol: Moxd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139296.2 | NP_647457.2 | 8 - 9 | 1207 | 97 | |
| GenBank mRNA | AB065134.1 | - | 8 - 9 | 1207 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Moxd2
Genomic Map