Assay Details
| Gene Symbol: Galnt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160404.1 | NP_001153876.1 | 11 - 12 | 1640 | 151 | |
| NM_013814.3 | NP_038842.3 | 11 - 12 | 1728 | 151 | ||
| GenBank mRNA | AK083391.1 | - | 11 - 12 | 1644 | 151 | |
| AK148550.1 | - | 5 - 6 | 708 | 151 | ||
| BC056215.1 | - | 11 - 12 | 1645 | 30019829 | 151 | |
| BC090962.1 | - | 10 - 11 | 1518 | 30626890 | 151 | |
| U73820.1 | - | 9 - 10 | 1393 | 151 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Galnt1
Genomic Map