Assay Details
| Gene Symbol: Bmx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009759.4 | NP_033889.2 | 1 - 2 | 149 | 126 | |
| GenBank mRNA | AK040936.1 | - | 1 - 2 | 150 | 126 | |
| AK080038.1 | - | 1 - 2 | 150 | 126 | ||
| U88091.1 | - | 1 - 2 | 135 | 126 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK040936.1, AK080038.1, NM_009759.4, U88091.1 transcripts.
Set Membership: |
|

Bmx
Genomic Map