Assay Details
| Gene Symbol: Bmx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009759.4 | NP_033889.2 | 9 - 10 | 969 | 69 | |
| GenBank mRNA | AF012104.1 | - | 8 - 9 | 822 | 69 | |
| AK036707.1 | - | 8 - 9 | 828 | 69 | ||
| AK040936.1 | - | 9 - 10 | 970 | 69 | ||
| AK080038.1 | - | 9 - 10 | 970 | 69 | ||
| BC138104.1 | - | 9 - 10 | 846 | 8861123 | 69 | |
| BC138105.1 | - | 9 - 10 | 846 | 8861124 | 69 | |
| U88091.1 | - | 9 - 10 | 955 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bmx
Genomic Map