Assay Details
| Gene Symbol: Nipal2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145469.5 | NP_663444.2 | 4 - 5 | 449 | 141 | |
| GenBank mRNA | AK033975.1 | - | 4 - 5 | 450 | 141 | |
| AK038556.1 | - | 4 - 5 | 590 | 141 | ||
| AK079085.1 | - | 4 - 5 | 882 | 141 | ||
| BC058207.1 | - | 4 - 5 | 615 | 5361417 | 141 | |
More Information
Set Membership: |
|

Nipal2
Genomic Map