Assay Details
| Gene Symbol: Nipal2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145469.5 | NP_663444.2 | 6 - 7 | 672 | 153 | |
| GenBank mRNA | AK033975.1 | - | 6 - 7 | 673 | 153 | |
| AK038556.1 | - | 6 - 7 | 814 | 154 | ||
| BC016107.1 | - | 6 - 7 | 695 | 4921508 | 153 | |
| BC030399.1 | - | 6 - 7 | 1189 | 4166825 | 153 | |
| BC058207.1 | - | 6 - 7 | 838 | 5361417 | 153 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nipal2
Genomic Map