Assay Details
| Gene Symbol: Cyp2r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177382.4 | NP_796356.2 | 2 - 3 | 381 | 90 | |
| GenBank mRNA | AY323818.1 | - | 2 - 3 | 381 | 90 | |
| BC108961.1 | - | 2 - 3 | 381 | 40046632 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cyp2r1
Genomic Map