Assay Details
| Gene Symbol: Cct5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007637.2 | NP_031663.1 | 10 - 11 | 1595 | 131 | |
| GenBank mRNA | AK077753.1 | - | 10 - 11 | 1554 | 131 | |
| AK088184.1 | - | 10 - 11 | 1529 | 131 | ||
| AK129056.1 | - | 10 - 11 | 1501 | 131 | ||
| AK133857.1 | - | 10 - 11 | 1553 | 131 | ||
| AK136493.1 | - | 10 - 11 | 1547 | 131 | ||
| AK145445.1 | - | 10 - 11 | 1564 | 131 | ||
| AK146405.1 | - | 10 - 11 | 1567 | 131 | ||
| AK146425.1 | - | 10 - 11 | 1547 | 131 | ||
| AK150390.1 | - | 10 - 11 | 1514 | 131 | ||
| AK153911.1 | - | 10 - 11 | 1571 | 131 | ||
| AK167895.1 | - | 10 - 11 | 1536 | 131 | ||
| AK213471.1 | - | 2 - 3 | 269 | 131 | ||
| AK216446.1 | - | 2 - 3 | 207 | 131 | ||
| AK219685.1 | - | 1 - 2 | 191 | 131 | ||
| BC092233.1 | - | 10 - 11 | 1565 | 132 | ||
| BC094427.1 | - | 10 - 11 | 1542 | 30537966 | 131 | |
| Z31555.1 | - | 10 - 11 | 1595 | 131 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30115243 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30115244 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30115242 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30115247 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30115246 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30115241 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30115248 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30115245 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30115249 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P144783 | BC092233.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P144783 | BC094427.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P144783 | Z31555.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P144783 | AK150390.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P144783 | AK167895.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P144783 | BC092233.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P144783 | BC094427.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P144783 | Z31555.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P144783 | AK150390.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P144783 | AK167895.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 425640 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 898394 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 309453 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 191300 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 821856 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 18443 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 159813 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P109516 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P178219 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P144783 | BC092233.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P144783 | BC094427.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P144783 | Z31555.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P144783 | AK150390.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P144783 | AK167895.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P144783 | BC092233.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P144783 | BC094427.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P144783 | Z31555.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P144783 | AK150390.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P144783 | AK167895.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P144783 | BC092233.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P144783 | BC094427.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P144783 | Z31555.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P144783 | AK150390.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P144783 | AK167895.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P144783 | BC092233.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P144783 | BC094427.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P144783 | Z31555.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P144783 | AK150390.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P144783 | AK167895.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30115228 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30115210 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30115214 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30115234 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30115242 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30115247 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30115235 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30115221 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30115222 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30115219 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30115216 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30115220 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30115229 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30115244 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30115248 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30115240 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30115217 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30115246 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30115212 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30115238 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30115231 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30115237 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30115227 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30115225 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30115230 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P178219 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P109516 | - | - |
| Whole Mouse Genome G4122A | A_51_P144783 | BC092233.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P144783 | BC094427.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P144783 | Z31555.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P144783 | AK150390.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P144783 | AK167895.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P144783 | BC092233.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P144783 | BC094427.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P144783 | Z31555.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P144783 | AK150390.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P144783 | AK167895.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139669 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106014 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P109516 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P178219 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11853836 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11853823 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11853827 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11853832 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30115241 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30115246 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30115243 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30115244 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30115245 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30115242 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30115249 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30115248 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30115247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115229 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115225 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115234 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115230 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115207 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115199 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115217 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12096296 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115209 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115227 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115240 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115231 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115242 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115221 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115235 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115216 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12159146 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115201 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115220 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115196 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12020513 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12096289 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115198 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115228 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115210 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115246 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115238 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115197 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12159151 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115212 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115219 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115214 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115247 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11853829 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10428055 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10428060 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10428062 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10428063 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10428054 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10428058 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10428064 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10428057 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10428065 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10428061 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10428059 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10428056 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417258_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10428052 | - | - |
| Mouse Genome 430 2.0 Array | 1417258_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417258_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12465_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12465_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500010287.mm.1 | NM_007637.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500010287.mm.1 | TC1500001326.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417258_at | - | - |
| Mouse Expression Set 430 | 1417258_PM_at | - | - |
| Murine Genome U74v2 Set | 93295_at | - | - |
| Murine Genome U74v2 Set | 161514_PM_at | - | - |
| Murine Genome U74v2 Set | 161514_at | - | - |
| Murine Genome U74v2 Set | 93295_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417258_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417258_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417258_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417258_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1050670 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1050670 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001050670 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2707308 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2707308 | - | - |
| mouse-6 v1.1 (Illumina) | scl47326.13.2_27-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1050670 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl47326.13.2_27-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1050670 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2707308 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2707308 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl47326.13.2_27-S | - | - |

Cct5
Genomic Map