Assay Details
| Gene Symbol: Sarnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025364.2 | NP_079640.1 | 3 - 4 | 350 | 88 | |
| GenBank mRNA | AK003453.1 | - | 3 - 4 | 202 | 88 | |
| AK018773.1 | - | 3 - 4 | 213 | 88 | ||
| AK088266.1 | - | 3 - 4 | 199 | 88 | ||
| AK146248.1 | - | 3 - 4 | 228 | 88 | ||
| AK212934.1 | - | 4 - 5 | 234 | 88 | ||
| AW824990.1 | - | 3 - 4 | 357 | 88 | ||
| BC027510.1 | - | 3 - 4 | 189 | 1382354 | 88 | |
| BC157909.1 | - | 3 - 4 | 214 | 9088111 | 88 | |
| BC158023.1 | - | 3 - 4 | 214 | 9007329 | 88 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sarnp
Genomic Map