Assay Details
| Gene Symbol: Rdh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134006.4 | NP_598767.1 | 1 - 2 | 151 | 65 | |
| GenBank mRNA | AF033196.2 | - | 1 - 2 | 117 | 65 | |
| AK135139.1 | - | 1 - 2 | 152 | 65 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF033196.2, AK135139.1, NM_134006.4 transcripts.
Set Membership: |
|

Rdh5
Genomic Map