Assay Details
| Gene Symbol: Scgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145399.1 | NP_663374.1 | 2 - 3 | 332 | 97 | |
| GenBank mRNA | AK148468.1 | - | 2 - 3 | 284 | 97 | |
| BC016093.1 | - | 2 - 3 | 332 | 4504330 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Scgn
Genomic Map