Assay Details
| Gene Symbol: Csn1s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286015.1 | NP_001272944.1 | 30 - 31 | 839 | 102 | |
| NM_007784.3 | NP_031810.1 | 31 - 32 | 863 | 102 | ||
| GenBank mRNA | AK085690.1 | - | 31 - 32 | 867 | 102 | |
| BC003859.1 | - | 31 - 32 | 847 | 3486384 | 102 | |
| BC006024.1 | - | 30 - 31 | 823 | 3592625 | 102 | |
| BC007192.1 | - | 31 - 32 | 847 | 102 | ||
| BC020321.1 | - | 12 - 13 | 317 | 102 | ||
| M36780.1 | - | 30 - 31 | 802 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Csn1s1
Genomic Map