Assay Details
| Gene Symbol: Zyx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289617.1 | NP_001276546.1 | 4 - 5 | 812 | 78 | |
| NM_011777.3 | NP_035907.1 | 4 - 5 | 812 | 78 | ||
| GenBank mRNA | AK031415.1 | - | 4 - 5 | 610 | 78 | |
| AK035743.1 | - | 4 - 5 | 583 | 78 | ||
| AK147812.1 | - | 4 - 5 | 813 | 78 | ||
| AK170571.1 | - | 4 - 5 | 586 | 78 | ||
| X99063.1 | - | 4 - 5 | 770 | 78 | ||
| Y07711.1 | - | 3 - 4 | 520 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zyx
Genomic Map