Assay Details
| Gene Symbol: 4930523O13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014957.1 | - | 6 - 7 | 896 | 104 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 1700022A22 gene | 1700022A22Rik | 4933413D07Rik | -68.2kb | Anti-sense | Chr. 15 - 46324585 - 46373899 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 4930548G14 gene | 4930548G14Rik | 170.2kb | Anti-sense | Chr. 15 - 46623305 - 46639858 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4930523O13Rik
Genomic Map