Assay Details
| Gene Symbol: Wdr86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081441.1 | NP_001074910.1 | 4 - 5 | 1194 | 118 | |
| GenBank mRNA | CN528185.1 | - | 2 - 3 | 500 | 118 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdr86
Genomic Map