Assay Details
| Gene Symbol: Gmds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146041.2 | NP_666153.1 | 9 - 10 | 1052 | 75 | |
| GenBank mRNA | BC031788.1 | - | 9 - 10 | 1052 | 4165509 | 75 |
| BC093502.1 | - | 9 - 10 | 1076 | 4215865 | 75 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gmds
Genomic Map