Assay Details
| Gene Symbol: Iqub | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172535.3 | NP_766123.2 | 11 - 12 | 2087 | 133 | |
| GenBank mRNA | AK029931.1 | - | 11 - 12 | 2077 | 133 | |
| AK133019.1 | - | 11 - 12 | 2069 | 133 | ||
| AK161501.1 | - | 5 - 6 | 912 | 133 | ||
| BC137937.1 | - | 11 - 12 | 2054 | 8860956 | 133 | |
| BC137957.1 | - | 11 - 12 | 2054 | 8860976 | 133 | |
More Information
Updated Mapping Information
Set Membership: |
|

Iqub
Genomic Map