Assay Details
| Gene Symbol: Grm8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008174.2 | NP_032200.2 | 5 - 6 | 1390 | 74 | |
| GenBank mRNA | AK038395.1 | - | 5 - 6 | 1444 | 74 | |
| AY673682.1 | - | 4 - 5 | 1018 | 74 | ||
| BC051384.1 | - | 5 - 6 | 1390 | 6466902 | 74 | |
| BC137596.1 | - | 4 - 5 | 1043 | 8860615 | 74 | |
| BC144753.1 | - | 4 - 5 | 1043 | 9053301 | 74 | |
| BC144754.1 | - | 4 - 5 | 1043 | 9053302 | 74 | |
| U17252.1 | - | 4 - 5 | 1088 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Grm8
Genomic Map