Assay Details
| Gene Symbol: Exoc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009148.3 | NP_033174.2 | 12 - 13 | 1873 | 127 | |
| GenBank mRNA | AF022962.1 | - | 12 - 13 | 1899 | 127 | |
| AK028250.1 | - | 10 - 11 | 1605 | 127 | ||
| AK173235.1 | - | 12 - 13 | 1876 | 127 | ||
| BC034644.1 | - | 12 - 13 | 1873 | 5355276 | 127 | |
More Information
Updated Mapping Information
Set Membership: |
|

Exoc4
Genomic Map