Assay Details
| Gene Symbol: Exoc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009148.3 | NP_033174.2 | 7 - 8 | 1184 | 75 | |
| GenBank mRNA | AF022962.1 | - | 7 - 8 | 1210 | 75 | |
| AK014499.1 | - | 7 - 8 | 1207 | 75 | ||
| AK036507.1 | - | 7 - 8 | 1196 | 75 | ||
| AK039983.1 | - | 7 - 8 | 1207 | 75 | ||
| AK086603.1 | - | 7 - 8 | 1203 | 75 | ||
| AK173235.1 | - | 7 - 8 | 1187 | 75 | ||
| AK200227.1 | - | 2 - 3 | 352 | 75 | ||
| BC034644.1 | - | 7 - 8 | 1184 | 5355276 | 75 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Exoc4
Genomic Map