Assay Details
| Gene Symbol: Arid4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194262.2 | NP_919238.1 | 18 - 19 | 2213 | 117 | |
| NM_198122.2 | NP_937755.1 | 17 - 18 | 1952 | 117 | ||
| GenBank mRNA | AK018194.1 | - | 1 - 2 | 3316 | 117 | |
| AK020165.1 | - | 3 - 4 | 350 | 117 | ||
| AK080967.1 | - | 2 - 3 | 114 | 117 | ||
| AK163022.1 | - | 1 - 2 | 702 | 117 | ||
| BC024724.1 | - | 3 - 4 | 491 | 117 | ||
| BC137783.1 | - | 17 - 18 | 1975 | 8860802 | 117 | |
| CB522383.1 | - | 4 - 5 | 628 | 117 | ||
More Information
Set Membership: |
|

Arid4b
Genomic Map