Assay Details
| Gene Symbol: Gmnn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020567.2 | NP_065592.1 | 2 - 3 | 182 | 112 | |
| GenBank mRNA | AF068780.1 | - | 2 - 3 | 329 | 112 | |
| AK131959.1 | - | 2 - 3 | 156 | 112 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF068780.1,AK131959.1,NM_020567.2 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gmnn
Genomic Map