Assay Details
| Gene Symbol: Myl6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172259.1 | NP_758463.1 | 6 - 7 | 695 | 69 | |
| GenBank mRNA | BC037527.1 | - | 6 - 7 | 695 | 3468485 | 69 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1952990 | NM_172259.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1952990 | BC037527.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 21374 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 67130 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 950538 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 207915 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 176611 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P110466 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1952990 | NM_172259.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1952990 | BC037527.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541716 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541708 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541710 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541696 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541723 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541688 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541691 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541689 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541721 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541713 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541725 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541693 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541717 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541705 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541704 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541700 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541697 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541736 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541727 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541720 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541730 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541690 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541728 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541692 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541722 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541719 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02247437 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P110466 | - | - |
| Whole Mouse Genome G4122A | A_52_P578692 | NM_172259.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P578692 | BC037527.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P476725 | AK017625.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116488 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112319 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P110466 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08475748 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08591010 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08591012 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08719263 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08719260 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08475750 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08719261 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08475745 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08591009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541693 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08283091 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541740 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541741 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541736 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541751 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541697 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541691 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541700 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541692 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541710 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541725 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08636057 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541743 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541721 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541753 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541719 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541727 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08827920 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541693 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08827917 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541744 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08827916 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08827907 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541755 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541722 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541690 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541704 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08878136 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541696 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541705 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541752 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08929837 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08827902 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541728 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541746 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08498003 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541708 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10373403 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373401 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373402 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373404 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373405 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373406 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430661_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10373400 | - | - |
| Mouse Genome 430 2.0 Array | 1430661_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1430661_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000023608.mm.1 | NM_172259.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023608.mm.1 | NM_172259.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023608.mm.1 | TC1000003146.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1430661_PM_at | - | - |
| Mouse Expression Set 430 | 1430661_at | - | - |
| Murine Genome U74v2 Set | 97101_at | - | - |
| Murine Genome U74v2 Set | 97101_PM_at | - | - |
| Murine Genome U74v2 Set | 92670_PM_at | - | - |
| Murine Genome U74v2 Set | 92670_at | - | - |
| Mouse Genome 430A 2.0 Array | 1430661_at | - | - |
| Mouse Genome 430A 2.0 Array | 1430661_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1430661_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1430661_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 468349 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6400546 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 101940452 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006400546 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2737183 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2737183 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1240281 | - | - |
| mouse-6 v1.1 (Illumina) | scl37360.7.1_32-S | - | - |
| mouse-6 v1.1 (Illumina) | scl38279.2_20-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 101940452 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6400546 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl37360.7.1_32-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6400546 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101940452 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2737183 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2737183 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1240281 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl37360.7.1_32-S | - | - |

Myl6b
Genomic Map