Assay Details
| Gene Symbol: Slc39a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028051.3 | NP_082327.2 | 1 - 2 | 238 | 91 | |
| GenBank mRNA | AK007473.1 | - | 1 - 2 | 239 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK007473.1, NM_028051.3 transcripts.
Set Membership: |
|

Slc39a5
Genomic Map